Assay Details
| Gene Symbol: Pnpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022601.2 | NP_072123.1 | 1 - 2 | 270 | 70 | |
| GenBank mRNA | BC087016.1 | - | 1 - 2 | 270 | 7128621 | 70 |
| U91561.1 | - | 1 - 2 | 261 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pnpo
Genomic Map