Assay Details
| Gene Symbol: Nat8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022635.1 | NP_072157.1 | 1 | 675 | 99 | |
| XM_006236789.2 | XP_006236851.1 | 2 | 757 | 99 | ||
| XM_006236790.2 | XP_006236852.1 | 2 | 767 | 99 | ||
| GenBank mRNA | AF185570.1 | - | 1 | 675 | 99 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nat8
Genomic Map