Assay Details
| Gene Symbol: Alpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022665.3 | NP_073156.3 | 1 - 2 | 113 | 86 | |
| XM_008767194.1 | XP_008765416.1 | 2 - 3 | 148 | 86 | ||
| GenBank mRNA | X17611.1 | - | 1 - 2 | 112 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Alpi
Genomic Map