Assay Details
| Gene Symbol: Fkbp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022675.1 | NP_073166.1 | 2 - 3 | 85 | 116 | |
| GenBank mRNA | D86642.1 | - | 2 - 3 | 85 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fkbp1b
Genomic Map