Assay Details
| Gene Symbol: Zfp238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022678.1 | NP_073169.1 | 1 | 649 | 68 | |
| XM_006250328.2 | XP_006250390.1 | 3 | 1742 | 68 | ||
| XM_006250330.2 | XP_006250392.1 | 2 | 1658 | 68 | ||
| XM_006250331.2 | XP_006250393.1 | 2 | 1137 | 68 | ||
| XM_008769796.1 | XP_008768018.1 | 2 | 2389 | 68 | ||
| XM_008769797.1 | XP_008768019.1 | 3 | 1521 | 68 | ||
| XM_008769798.1 | XP_008768020.1 | 3 | 1741 | 68 | ||
| XM_008769799.1 | XP_008768021.1 | 2 | 3660 | 68 | ||
| XM_008769800.1 | XP_008768022.1 | 2 | 692 | 68 | ||
| GenBank mRNA | AF221838.1 | - | 1 | 649 | 68 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp238
Genomic Map