Assay Details
| Gene Symbol: Pde1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022710.1 | NP_073201.1 | 6 - 7 | 661 | 65 | |
| XM_006242386.2 | XP_006242448.1 | 5 - 6 | 1233 | 65 | ||
| GenBank mRNA | AF327906.1 | - | 6 - 7 | 654 | 65 | |
| FQ212460.1 | - | 6 - 7 | 717 | 65 | ||
| M94537.1 | - | 6 - 7 | 661 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pde1b
Genomic Map