Assay Details
| Gene Symbol: Kcnmb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023960.2 | NP_076450.1 | 1 - 2 | 354 | 79 | |
| GenBank mRNA | AB050637.1 | - | 1 - 2 | 336 | 79 | |
| AY028605.1 | - | 1 - 2 | 354 | 79 | ||
| BC161829.1 | - | 1 - 2 | 534 | 9033053 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnmb4
Genomic Map