Assay Details
| Gene Symbol: Hivep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024137.1 | NP_077051.1 | 5 - 6 | 6254 | 82 | |
| GenBank mRNA | D37951.1 | - | 5 - 6 | 6254 | 82 | |
| M65251.1 | - | 3 - 4 | 927 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P146821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649322 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916925 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276351 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70218 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130179 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112756 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P146821 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P146821 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P146821 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P146821 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701288823 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49939 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 2254 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 77009 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23011 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 40818 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22872 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 72698 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 34186 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103785 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701288823 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10701836 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10701837 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10701831 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10701834 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10701838 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10701833 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10701835 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10701832 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10701830 | - | - |
| Rat Genome 230 2.0 Array | 1368546_at | - | - |
| Rat Genome 230 2.0 Array | 1368546_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368546_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368546_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100000431.rn.1 | TC0100000093.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100000432.rn.1 | TC0100000093.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368546_PM_at | - | - |
| Rat Expression Set 230 | 1368546_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10701830 | - | - |
| Rat Genome U34 Set | M65251_s_at | - | - |
| Rat Genome U34 Set | D37951UTR#1_at | - | - |
| Rat Genome U34 Set | D37951cds_s_at | - | - |
| Rat Genome U34 Set | X54249mRNA_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21376566 | - | - |

Hivep2
Genomic Map