Assay Details
| Gene Symbol: Mepe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024142.1 | NP_077056.1 | 1 - 2 | 49 | 67 | |
| GenBank mRNA | AF260922.1 | - | 1 - 2 | 49 | 67 | |
| AF530558.1 | - | 1 - 2 | 46 | 67 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF260922.1,AF530558.1,NM_024142.1 transcripts.
Set Membership: |
|

Mepe
Genomic Map