Assay Details
| Gene Symbol: Gpr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037295.1 | NP_001032372.1 | 2 | 1281 | 83 | |
| NM_030831.1 | NP_110458.1 | 1 | 1451 | 83 | ||
| GenBank mRNA | BC127447.1 | - | 2 | 1209 | 8363428 | 83 |
| U12184.1 | - | 1 | 1451 | 83 | ||
| X61496.1 | - | 2 | 1279 | 84 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gpr12
Genomic Map