Assay Details
| Gene Symbol: Bdkrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030851.1 | NP_110478.1 | 1 - 2 | 86 | 105 | |
| GenBank mRNA | AJ132230.2 | - | 1 - 2 | 86 | 105 | |
| U66107.2 | - | 1 - 2 | 95 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ132230.2, NM_030851.1, U66107.2 transcripts.
Set Membership: |
|

Bdkrb1
Genomic Map