Assay Details
| Gene Symbol: Marcksl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030862.2 | NP_110489.1 | 2 | 662 | 93 | |
| GenBank mRNA | AJ301677.1 | - | 2 | 555 | 93 | |
| BC081789.1 | - | 2 | 662 | 7134056 | 93 | |
| Gene Symbol: LOC102550530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759283.1 | XP_008757505.1 | 2 | 621 | 93 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Marcksl1
Genomic Map