Assay Details
| Gene Symbol: Sh3gl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031239.2 | NP_112518.1 | 8 - 9 | 986 | 105 | |
| GenBank mRNA | AB008161.1 | - | 8 - 9 | 986 | 105 | |
| AF009602.1 | - | 8 - 9 | 971 | 105 | ||
| BC070893.1 | - | 8 - 9 | 982 | 7099223 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sh3gl1
Genomic Map