Assay Details
| Gene Symbol: Slc6a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031343.1 | NP_112633.1 | 3 - 4 | 686 | 109 | |
| GenBank mRNA | AB021970.1 | - | 3 - 4 | 686 | 109 | |
| AB021971.1 | - | 3 - 4 | 686 | 109 | ||
| Y13223.1 | - | 3 - 4 | 644 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
neurotransmitter transport
monoamine transport
norepinephrine transport
response to drug
response to pain
transmembrane transport
neurotransmitter:sodium symporter activity
norepinephrine transmembrane transporter activity
protein binding
monoamine transmembrane transporter activity
alpha-tubulin binding
beta-tubulin binding
neurotransmitter transport
monoamine transport
norepinephrine transport
response to drug
response to pain
transmembrane transport
neurotransmitter:sodium symporter activity
norepinephrine transmembrane transporter activity
protein binding
monoamine transmembrane transporter activity
alpha-tubulin binding
beta-tubulin binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P146767 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P15317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99410 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497769 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55501 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101426 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P15317 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P146767 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P15317 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P146767 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P15317 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P146767 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P304202 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18100 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 2346 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38580 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10809520 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809510 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809516 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809511 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809517 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809513 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809509 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809515 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809508 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809518 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809512 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809521 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809514 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809519 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809522 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10809507 | - | - |
| Rat Genome 230 2.0 Array | 1393399_at | - | - |
| Rat Genome 230 2.0 Array | 1387418_PM_a_at | - | - |
| Rat Genome 230 2.0 Array | 1387418_a_at | - | - |
| Rat Genome 230 2.0 Array | 1393399_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387418_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1387418_PM_a_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1900008398.rn.1 | TC1900001050.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900008400.rn.1 | TC1900001050.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900008404.rn.1 | TC1900001050.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900008392.rn.1 | TC1900001050.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900008396.rn.1 | TC1900001050.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900008403.rn.1 | TC1900001050.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900008393.rn.1 | TC1900001050.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900008385.rn.1 | TC1900001050.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900008386.rn.1 | TC1900001050.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900008397.rn.1 | TC1900001050.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900008402.rn.1 | TC1900001050.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900008394.rn.1 | TC1900001050.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900008395.rn.1 | TC1900001050.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900008399.rn.1 | TC1900001050.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900008401.rn.1 | TC1900001050.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387418_PM_a_at | - | - |
| Rat Expression Set 230 | 1387418_a_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809507 | - | - |
| Rat Genome U34 Set | Y13223cds_at | - | - |
| Rat Genome U34 Set | L29573_s_at | - | - |
| Rat Genome U34 Set | rc_AI170447_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20853329 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1362601 | - | - |
| RatRef-12 Expression BeadChip | ILMN_70414 | - | - |

Slc6a2
Genomic Map