Assay Details
| Gene Symbol: Cd200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031518.2 | NP_113706.2 | 1 - 2 | 80 | 59 | |
| GenBank mRNA | BC089793.1 | - | 1 - 2 | 80 | 7380792 | 59 |
| X01785.1 | - | 1 - 2 | 37 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC089793.1, NM_031518.2, X01785.1 transcripts.
Set Membership: |
|

Cd200
Genomic Map