Assay Details
| Gene Symbol: Wt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031534.2 | NP_113722.2 | 7 - 8 | 1060 | 104 | |
| XM_006234620.1 | XP_006234682.1 | 6 - 7 | 1402 | 104 | ||
| XM_006234621.2 | XP_006234683.1 | 7 - 8 | 1531 | 104 | ||
| XM_006234622.2 | XP_006234684.1 | 6 - 7 | 1480 | 104 | ||
| GenBank mRNA | X69716.1 | - | 7 - 8 | 1060 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
vasculogenesis
metanephros development
ureteric bud development
branching involved in ureteric bud morphogenesis
kidney development
mesonephros development
regulation of organ formation
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
apoptotic process
germ cell development
spermatogenesis
single fertilization
thorax and anterior abdomen determination
heart development
sex determination
negative regulation of cell proliferation
RNA splicing
gonad development
male gonad development
tissue development
regulation of gene expression
negative regulation of translation
negative regulation of cell growth
sperm motility
adrenal gland development
male genitalia development
epithelial cell differentiation
glomerulus development
glomerular basement membrane development
adrenal cortex formation
camera-type eye development
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
Sertoli cell development
mesenchymal to epithelial transition
positive regulation of heart growth
diaphragm development
cardiac muscle cell fate commitment
visceral serous pericardium development
cellular response to cAMP
cellular response to gonadotropin stimulus
metanephric mesenchyme development
glomerular visceral epithelial cell differentiation
posterior mesonephric tubule development
metanephric epithelium development
metanephric S-shaped body morphogenesis
negative regulation of metanephric glomerular mesangial cell proliferation
seminiferous tubule development
negative regulation of mesenchymal cell apoptotic process involved in metanephros development
positive regulation of male gonad development
negative regulation of female gonad development
positive regulation of metanephric ureteric bud development
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
nucleic acid binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
RNA binding
protein binding
zinc ion binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding
C2H2 zinc finger domain binding
negative regulation of transcription from RNA polymerase II promoter
vasculogenesis
metanephros development
ureteric bud development
branching involved in ureteric bud morphogenesis
kidney development
mesonephros development
regulation of organ formation
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
apoptotic process
germ cell development
spermatogenesis
single fertilization
thorax and anterior abdomen determination
heart development
sex determination
negative regulation of cell proliferation
RNA splicing
gonad development
male gonad development
tissue development
regulation of gene expression
negative regulation of translation
negative regulation of cell growth
sperm motility
adrenal gland development
male genitalia development
epithelial cell differentiation
glomerulus development
glomerular basement membrane development
adrenal cortex formation
camera-type eye development
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
Sertoli cell development
mesenchymal to epithelial transition
positive regulation of heart growth
diaphragm development
cardiac muscle cell fate commitment
visceral serous pericardium development
cellular response to cAMP
cellular response to gonadotropin stimulus
metanephric mesenchyme development
glomerular visceral epithelial cell differentiation
posterior mesonephric tubule development
metanephric epithelium development
metanephric S-shaped body morphogenesis
negative regulation of metanephric glomerular mesangial cell proliferation
seminiferous tubule development
negative regulation of mesenchymal cell apoptotic process involved in metanephros development
positive regulation of male gonad development
negative regulation of female gonad development
positive regulation of metanephric ureteric bud development
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
nucleic acid binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
RNA binding
protein binding
zinc ion binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding
C2H2 zinc finger domain binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P142861 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P140507 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P808666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152907 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P140507 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P808666 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P142861 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P142861 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P808666 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P140507 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P808666 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P140507 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P142861 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P176466 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 43840 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68410 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 37941 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10838248 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838244 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838253 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838252 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838247 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838250 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838246 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838249 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838245 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838251 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838254 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10838242 | - | - |
| Rat Genome 230 2.0 Array | 1377223_at | - | - |
| Rat Genome 230 2.0 Array | 1369695_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1377223_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1369695_at | - | - |
| Affymetrix HT Rat Focus Array | 1369695_at | - | - |
| Affymetrix HT Rat Focus Array | 1369695_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300008968.rn.1 | TC0300001321.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369695_at | - | - |
| Rat Expression Set 230 | 1377223_PM_at | - | - |
| Rat Expression Set 230 | 1369695_PM_at | - | - |
| Rat Expression Set 230 | 1377223_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838242 | - | - |
| Rat Genome U34 Set | rc_AA965119_at | - | - |
| Rat Genome U34 Set | S63358_s_at | - | - |
| Rat Genome U34 Set | X69716_at | - | - |
| Rat Genome U34 Set | rc_AA899753_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22175193 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1360850 | - | - |
| RatRef-12 Expression BeadChip | ILMN_56820 | - | - |

Wt1
Genomic Map