Assay Details
| Gene Symbol: Clcn7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031568.2 | NP_113756.1 | 17 - 18 | 1681 | 62 | |
| GenBank mRNA | Z67744.1 | - | 17 - 18 | 1682 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P010363 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888596 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162570 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P010363 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P010363 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P010363 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12590 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701195752 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 81062 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14541 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10915 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701195752 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10732313 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732308 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732318 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732310 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732311 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732305 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732319 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732322 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732307 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732328 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732303 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732317 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732306 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732309 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732321 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732314 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732326 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732315 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732304 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732323 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732325 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732324 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732312 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732316 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732327 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10732302 | - | - |
| Rat Genome 230 2.0 Array | 1388509_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388509_at | - | - |
| Rat Genome 230 2.0 Array | 1369700_at | - | - |
| Rat Genome 230 2.0 Array | 1369700_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369700_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369700_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000002329.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002337.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002352.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002333.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002353.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002339.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002348.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002351.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002355.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002336.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002350.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002331.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002346.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002357.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002328.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002332.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002338.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002343.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002356.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002334.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002340.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002341.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002342.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002345.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002330.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002335.rn.1 | TC1000000235.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000002344.rn.1 | TC1000000235.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369700_PM_at | - | - |
| Rat Expression Set 230 | 1369700_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732302 | - | - |
| Rat Genome U34 Set | Z67744_at | - | - |
| Rat Genome U34 Set | Z67744_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20865309 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365342 | - | - |
| RatRef-12 Expression BeadChip | ILMN_53810 | - | - |

Clcn7
Genomic Map