Assay Details
| Gene Symbol: Capn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031673.2 | NP_113861.1 | 266 | 56 | ||
| GenBank mRNA | AB113362.1 | - | 173 | 56 | ||
| AB113363.1 | - | 173 | 56 | |||
| AF227909.3 | - | 166 | 56 | |||
| BC085725.1 | - | 266 | 7123652 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Capn10
Genomic Map