Assay Details
| Gene Symbol: Slc16a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031744.2 | NP_113932.2 | 1 - 2 | 117 | 91 | |
| XM_006242039.2 | XP_006242101.1 | 1 - 2 | 246 | 91 | ||
| GenBank mRNA | AF059258.1 | - | 1 - 2 | 117 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF059258.1, NM_031744.2, XM_006242039.2 transcripts.
Set Membership: |
|

Slc16a8
Genomic Map