Assay Details
| Gene Symbol: Galnt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031796.1 | NP_113984.1 | 2 - 3 | 1703 | 88 | |
| GenBank mRNA | AF049344.1 | - | 2 - 3 | 1703 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P325829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661586 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555520 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879921 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90013 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P325829 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P325829 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P325829 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P325829 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 32336 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38279 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19731 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 33922 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 64981 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10836300 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10836307 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10836305 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10836306 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10836301 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10836302 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10836304 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10836299 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10836303 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10836308 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10836298 | - | - |
| Rat Genome 230 2.0 Array | 1387554_at | - | - |
| Rat Genome 230 2.0 Array | 1387554_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387554_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387554_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300004802.rn.1 | TC0300000625.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387554_PM_at | - | - |
| Rat Expression Set 230 | 1387554_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10836298 | - | - |
| Rat Genome U34 Set | AF049344_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21965695 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349069 | - | - |
| RatRef-12 Expression BeadChip | ILMN_65992 | - | - |

Galnt5
Genomic Map