Assay Details
| Gene Symbol: Plk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031821.1 | NP_114009.1 | 1 - 2 | 392 | 138 | |
| GenBank mRNA | AF136583.1 | - | 1 - 2 | 392 | 138 | |
| BC070878.1 | - | 1 - 2 | 376 | 7096750 | 138 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plk2
Genomic Map