Assay Details
| Gene Symbol: Trpv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031982.1 | NP_114188.1 | 15 - 16 | 2422 | 66 | |
| XM_008767882.1 | XP_008766104.1 | 14 - 15 | 2429 | 66 | ||
| GenBank mRNA | AB040873.1 | - | 14 - 15 | 2342 | 66 | |
| AB041029.1 | - | 13 - 14 | 2162 | 66 | ||
| AF029310.1 | - | 15 - 16 | 2422 | 66 | ||
| AF158248.2 | - | 9 - 10 | 1755 | 66 | ||
| LC008303.1 | - | 10 - 11 | 1662 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
temperature homeostasis
fever generation
microglial cell activation
diet induced thermogenesis
peptide secretion
lipid metabolic process
calcium ion transport
inflammatory response
signal transduction
positive regulation of cytosolic calcium ion concentration
response to pH
response to heat
response to organonitrogen compound
glutamate secretion
urinary bladder smooth muscle contraction
sensory perception of pain
ion transmembrane transport
positive regulation of apoptotic process
response to peptide hormone
positive regulation of nitric oxide biosynthetic process
response to pain
behavioral response to pain
sensory perception of mechanical stimulus
thermoception
detection of chemical stimulus involved in sensory perception of pain
smooth muscle contraction involved in micturition
positive regulation of gastric acid secretion
calcium ion transmembrane transport
cellular response to alkaloid
cellular response to ATP
cellular response to cytokine stimulus
cellular response to tumor necrosis factor
cellular response to growth factor stimulus
cellular response to temperature stimulus
negative regulation of establishment of blood-brain barrier
cellular response to nerve growth factor stimulus
transmembrane signaling receptor activity
ion channel activity
excitatory extracellular ligand-gated ion channel activity
cation channel activity
calcium channel activity
protein binding
calmodulin binding
ATP binding
cation transmembrane transporter activity
ligand-gated ion channel activity
calcium-release channel activity
chloride channel regulator activity
identical protein binding
phosphoprotein binding
temperature homeostasis
fever generation
microglial cell activation
diet induced thermogenesis
peptide secretion
lipid metabolic process
calcium ion transport
inflammatory response
signal transduction
positive regulation of cytosolic calcium ion concentration
response to pH
response to heat
response to organonitrogen compound
glutamate secretion
urinary bladder smooth muscle contraction
sensory perception of pain
ion transmembrane transport
positive regulation of apoptotic process
response to peptide hormone
positive regulation of nitric oxide biosynthetic process
response to pain
behavioral response to pain
sensory perception of mechanical stimulus
thermoception
detection of chemical stimulus involved in sensory perception of pain
smooth muscle contraction involved in micturition
positive regulation of gastric acid secretion
calcium ion transmembrane transport
cellular response to alkaloid
cellular response to ATP
cellular response to cytokine stimulus
cellular response to tumor necrosis factor
cellular response to growth factor stimulus
cellular response to temperature stimulus
negative regulation of establishment of blood-brain barrier
cellular response to nerve growth factor stimulus
transmembrane signaling receptor activity
ion channel activity
excitatory extracellular ligand-gated ion channel activity
cation channel activity
calcium channel activity
protein binding
calmodulin binding
ATP binding
cation transmembrane transporter activity
ligand-gated ion channel activity
calcium-release channel activity
chloride channel regulator activity
identical protein binding
phosphoprotein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P130201 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P061740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440085 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140668 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P130201 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P061740 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P130201 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P061740 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P130201 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P061740 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12709 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701821845 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 51631 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 52817 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701821845 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10735682 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10735687 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10735680 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10735691 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10735697 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759110 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10735696 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10735694 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10735688 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10735686 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10735695 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10735684 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759111 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10735690 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10735681 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10735679 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10735698 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10735692 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10735678 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10735685 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10735693 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10735683 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759112 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10735689 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10759096 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10735677 | - | - |
| Rat Genome 230 2.0 Array | 1369370_PM_s_at | - | - |
| Rat Genome 230 2.0 Array | 1370685_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368313_PM_a_at | - | - |
| Rat Genome 230 2.0 Array | 1370675_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1369370_s_at | - | - |
| Rat Genome 230 2.0 Array | 1370675_at | - | - |
| Rat Genome 230 2.0 Array | 1368313_a_at | - | - |
| Rat Genome 230 2.0 Array | 1370685_at | - | - |
| Affymetrix HT Rat Focus Array | 1370675_at | - | - |
| Affymetrix HT Rat Focus Array | 1368313_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1370685_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370685_at | - | - |
| Affymetrix HT Rat Focus Array | 1370675_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368313_PM_a_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000009387.rn.1 | TC1000001025.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000009385.rn.1 | TC1000001025.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369370_s_at | - | - |
| Rat Expression Set 230 | 1369370_PM_s_at | - | - |
| Rat Expression Set 230 | 1370685_at | - | - |
| Rat Expression Set 230 | 1370675_PM_at | - | - |
| Rat Expression Set 230 | 1370685_PM_at | - | - |
| Rat Expression Set 230 | 1368313_a_at | - | - |
| Rat Expression Set 230 | 1368313_PM_a_at | - | - |
| Rat Expression Set 230 | 1370675_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10735677 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759096 | - | - |
| Rat Genome U34 Set | AF029310_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21426000 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1356230 | - | - |
| RatRef-12 Expression BeadChip | ILMN_65825 | - | - |

Trpv1
Genomic Map