Assay Details
| Gene Symbol: Gstt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053293.2 | NP_445745.1 | 2 - 3 | 229 | 83 | |
| XM_008772860.1 | XP_008771082.1 | 2 - 3 | 726 | 83 | ||
| GenBank mRNA | BC086426.1 | - | 2 - 3 | 229 | 7301492 | 83 |
| FJ179408.1 | - | 2 - 3 | 199 | 83 | ||
| X67654.1 | - | 2 - 3 | 199 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gstt1
Genomic Map