Assay Details
| Gene Symbol: LOC100910732 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751066.2 | XP_003751114.1 | 13 - 14 | 1627 | 68 | |
| XM_006248473.1 | XP_006248535.1 | 14 - 15 | 1756 | 68 | ||
| XM_008768778.1 | XP_008767000.1 | 13 - 14 | 3188 | 68 | ||
| Gene Symbol: Pak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053306.3 | NP_445758.2 | 1479 | 68 | ||
| GenBank mRNA | S80221.1 | - | 1398 | 68 | ||
| U19967.1 | - | 1479 | 68 | |||
| U35345.1 | - | 1398 | 68 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100910732
Genomic Map