Assay Details
| Gene Symbol: Cxcr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053415.1 | NP_445867.1 | 1 - 2 | 165 | 58 | |
| GenBank mRNA | AF223642.1 | - | 1 - 2 | 165 | 58 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF223642.1, NM_053415.1 transcripts.
Set Membership: |
|

Cxcr3
Genomic Map