Assay Details
| Gene Symbol: Ddx39a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006255314.2 | XP_006255376.1 | 1 - 2 | 202 | 81 | |
| GenBank mRNA | AF063447.1 | - | 1 - 2 | 98 | 81 | |
| FQ231639.1 | - | 1 - 2 | 207 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF063447.1, XM_006255314.2 transcripts.
Set Membership: |
|

Ddx39a
Genomic Map