Assay Details
| Gene Symbol: Ece1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053596.2 | NP_446048.2 | 4 - 5 | 506 | 86 | |
| XM_006239275.2 | XP_006239337.1 | 4 - 5 | 1094 | 86 | ||
| XM_006239276.2 | XP_006239338.1 | 3 - 4 | 1110 | 86 | ||
| GenBank mRNA | BC072504.1 | - | 4 - 5 | 506 | 7099038 | 86 |
| D29683.1 | - | 4 - 5 | 541 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
positive regulation of receptor recycling
proteolysis
apoptotic process
heart development
regulation of blood pressure
positive regulation of cardiac muscle hypertrophy
substance P catabolic process
bradykinin catabolic process
calcitonin catabolic process
protein processing
peptide hormone processing
positive regulation of cAMP biosynthetic process
endothelin maturation
response to muscle stretch
regulation of vasodilation
hormone catabolic process
response to drug
embryonic digit morphogenesis
ear development
positive regulation of G-protein coupled receptor protein signaling pathway
response to cadmium ion
pharyngeal system development
regulation of ERK1 and ERK2 cascade
endopeptidase activity
metalloendopeptidase activity
protein homodimerization activity
metal ion binding
response to hypoxia
positive regulation of receptor recycling
proteolysis
apoptotic process
heart development
regulation of blood pressure
positive regulation of cardiac muscle hypertrophy
substance P catabolic process
bradykinin catabolic process
calcitonin catabolic process
protein processing
peptide hormone processing
positive regulation of cAMP biosynthetic process
endothelin maturation
response to muscle stretch
regulation of vasodilation
hormone catabolic process
response to drug
embryonic digit morphogenesis
ear development
positive regulation of G-protein coupled receptor protein signaling pathway
response to cadmium ion
pharyngeal system development
regulation of ERK1 and ERK2 cascade
endopeptidase activity
metalloendopeptidase activity
protein homodimerization activity
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P839121 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P991864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925351 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603834 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763798 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13186 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P839121 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P991864 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P991864 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P839121 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P991864 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P839121 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P991864 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P839121 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P195356 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701276882 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700179153 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 39548 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 98224 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5996 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 45539 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 98899 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6887 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 41219 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 43341 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701276882 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700179153 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10873221 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873223 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873206 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873210 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873209 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873218 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873216 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873215 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873208 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873213 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873212 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873207 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873225 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873204 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873222 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873220 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873217 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873224 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873214 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873219 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873211 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10873203 | - | - |
| Rat Genome 230 2.0 Array | 1367801_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1367801_at | - | - |
| Affymetrix HT Rat Focus Array | 1367801_at | - | - |
| Affymetrix HT Rat Focus Array | 1367801_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500012786.rn.1 | TC0500001734.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500012785.rn.1 | TC0500001734.rn.1 | RTA TC |
| Rat Expression Set 230 | 1367801_at | - | - |
| Rat Expression Set 230 | 1367801_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10873203 | - | - |
| Rat Genome U34 Set | D29683_at | - | - |
| Rat Genome U34 Set | rc_AA956930_s_at | - | - |
| Rat Genome U34 Set | rc_AA900686_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21416209 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358385 | - | - |
| RatRef-12 Expression BeadChip | ILMN_51504 | - | - |

Ece1
Genomic Map