Assay Details
| Gene Symbol: Gfm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053625.1 | NP_446077.1 | 13 - 14 | 1741 | 115 | |
| GenBank mRNA | BC085721.1 | - | 13 - 14 | 1695 | 7192461 | 113 |
| L14684.1 | - | 13 - 14 | 1741 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P038958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479037 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466285 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175896 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68645 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128818 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P038958 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P038958 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P038958 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P296956 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 61878 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 74302 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46704 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 39976 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10815817 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815825 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815831 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815826 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815824 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815830 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815833 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815816 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815821 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815822 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815832 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815820 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815818 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815819 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815829 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815828 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815827 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815823 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10815815 | - | - |
| Rat Genome 230 2.0 Array | 1367756_at | - | - |
| Rat Genome 230 2.0 Array | 1367756_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1367756_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1367756_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200008386.rn.1 | TC0200001345.rn.1 | RTA TC |
| Rat Expression Set 230 | 1367756_PM_at | - | - |
| Rat Expression Set 230 | 1367756_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815815 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21519503 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376764 | - | - |

Gfm1
Genomic Map