Assay Details
| Gene Symbol: Ltc4s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053639.2 | NP_446091.2 | 1 - 2 | 83 | 94 | |
| XM_008767655.1 | XP_008765877.1 | 2 - 3 | 1104 | 94 | ||
| GenBank mRNA | AB048790.2 | - | 1 - 2 | 83 | 94 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB048790.2,NM_053639.2,XM_008767655.1 transcripts.
Set Membership: |
|

Ltc4s
Genomic Map