Assay Details
| Gene Symbol: Ltb4r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053640.1 | NP_446092.1 | 1 | 1038 | 103 | |
| XM_008770658.1 | XP_008768880.1 | 1 | 1058 | 103 | ||
| GenBank mRNA | AB052660.1 | - | 1 | 1038 | 103 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ltb4r2
Genomic Map