Assay Details
| Gene Symbol: Cysltr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053641.1 | NP_446093.1 | 1 | 356 | 97 | |
| XM_006257158.2 | XP_006257220.1 | 3 | 1070 | 97 | ||
| GenBank mRNA | AB052685.1 | - | 1 | 356 | 97 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cysltr1
Genomic Map