Assay Details
| Gene Symbol: Dmrt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053706.1 | NP_446158.1 | 4 - 5 | 1063 | 76 | |
| GenBank mRNA | AF379608.1 | - | 4 - 5 | 1063 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
cell morphogenesis
male germ cell proliferation
developmental process involved in reproduction
transcription from RNA polymerase II promoter
spermatogenesis
germ cell migration
male gonad development
male sex determination
intracellular signal transduction
negative regulation of meiotic nuclear division
positive regulation of mitotic nuclear division
positive regulation of transcription from RNA polymerase II promoter
male sex differentiation
oocyte development
Sertoli cell differentiation
Sertoli cell development
positive regulation of meiosis I
regulation of nodal signaling pathway
positive regulation of male gonad development
RNA polymerase II regulatory region sequence-specific DNA binding
core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein homodimerization activity
sequence-specific DNA binding
protein heterodimerization activity
negative regulation of transcription from RNA polymerase II promoter
cell morphogenesis
male germ cell proliferation
developmental process involved in reproduction
transcription from RNA polymerase II promoter
spermatogenesis
germ cell migration
male gonad development
male sex determination
intracellular signal transduction
negative regulation of meiotic nuclear division
positive regulation of mitotic nuclear division
positive regulation of transcription from RNA polymerase II promoter
male sex differentiation
oocyte development
Sertoli cell differentiation
Sertoli cell development
positive regulation of meiosis I
regulation of nodal signaling pathway
positive regulation of male gonad development
RNA polymerase II regulatory region sequence-specific DNA binding
core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein homodimerization activity
sequence-specific DNA binding
protein heterodimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P092614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73891 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720352 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189895 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254369 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695140 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920531 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883403 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197177 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104385 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P092614 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P092614 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P092614 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P149376 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14714 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 8681 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 74951 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19559 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6212 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49627 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 40659 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10714522 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10714520 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10714521 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10714523 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10714519 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10714518 | - | - |
| Rat Genome 230 2.0 Array | 1387597_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387597_at | - | - |
| Affymetrix HT Rat Focus Array | 1387597_at | - | - |
| Affymetrix HT Rat Focus Array | 1387597_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100027675.rn.1 | TC0100003406.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387597_PM_at | - | - |
| Rat Expression Set 230 | 1387597_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10714518 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21979087 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368167 | - | - |
| RatRef-12 Expression BeadChip | ILMN_64327 | - | - |

Dmrt1
Genomic Map