Assay Details
| Gene Symbol: Rap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053741.1 | NP_446193.1 | 1 | 351 | 106 | |
| GenBank mRNA | AY037826.1 | - | 1 | 351 | 106 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rap2a
Genomic Map