Assay Details
| Gene Symbol: Lim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053771.1 | NP_446223.1 | 4 - 5 | 506 | 55 | |
| GenBank mRNA | L04191.1 | - | 4 - 5 | 484 | 55 | |
| M87053.1 | - | 4 - 5 | 506 | 55 | ||
| S55224.1 | - | 4 - 5 | 497 | 55 | ||
More Information
Set Membership: |
|

Lim2
Genomic Map