Assay Details
| Gene Symbol: Sacm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053798.2 | NP_446250.1 | 14 - 15 | 1275 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P226461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351858 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84140 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P226461 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P226461 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P226461 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P226461 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 43790 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 43399 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31837 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10914599 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914594 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914596 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914592 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914584 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914595 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914597 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914588 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914589 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914586 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914591 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914593 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914585 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914590 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914587 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914600 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914582 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914581 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914583 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914598 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10914580 | - | - |
| Rat Genome 230 2.0 Array | 1368433_at | - | - |
| Rat Genome 230 2.0 Array | 1368433_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368433_at | - | - |
| Affymetrix HT Rat Focus Array | 1368433_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800014771.rn.1 | TC0800001840.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800014770.rn.1 | TC0800001840.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368433_PM_at | - | - |
| Rat Expression Set 230 | 1368433_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914580 | - | - |
| Rat Genome U34 Set | rc_AI177608_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21813611 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376512 | - | - |

Sacm1l
Genomic Map