Assay Details
| Gene Symbol: Timp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053819.1 | NP_446271.1 | 2 - 3 | 154 | 91 | |
| XM_006256608.2 | XP_006256670.1 | 2 - 3 | 218 | 91 | ||
| XM_008773034.1 | XP_008771256.1 | 1 - 2 | 128 | 91 | ||
| GenBank mRNA | AY550026.1 | - | 1 - 2 | 143 | 91 | |
| BC099821.1 | - | 2 - 3 | 152 | 7930800 | 91 | |
| FQ221763.1 | - | 2 - 3 | 164 | 91 | ||
| FQ222271.1 | - | 2 - 3 | 164 | 91 | ||
| FQ222437.1 | - | 2 - 3 | 167 | 91 | ||
| FQ222915.1 | - | 2 - 3 | 164 | 91 | ||
| FQ228400.1 | - | 2 - 3 | 163 | 91 | ||
| FQ230039.1 | - | 2 - 3 | 161 | 91 | ||
| L31883.1 | - | 2 - 3 | 154 | 91 | ||
| U06179.1 | - | 1 - 2 | 137 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Timp1
Genomic Map