Assay Details
| Gene Symbol: Plod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053827.2 | NP_446279.2 | 18 - 19 | 2076 | 77 | |
| GenBank mRNA | CV080837.1 | - | 6 - 7 | 579 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P306355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341520 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178400 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P306355 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P306355 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P306355 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15345 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P306355 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701577909 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 97879 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17435 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 77362 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701577909 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10881540 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881522 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881529 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881533 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881523 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881525 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742970 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881537 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881526 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881528 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881530 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881541 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881535 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881536 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881534 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742969 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881531 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881532 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881524 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881527 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881539 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10742968 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10881521 | - | - |
| Rat Genome 230 2.0 Array | 1392873_at | - | - |
| Rat Genome 230 2.0 Array | 1392873_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1367807_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1367807_at | - | - |
| Affymetrix HT Rat Focus Array | 1367807_at | - | - |
| Affymetrix HT Rat Focus Array | 1367807_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500030680.rn.1 | TC0500004033.rn.1 | RTA TC |
| Rat Expression Set 230 | 1367807_at | - | - |
| Rat Expression Set 230 | 1367807_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881521 | - | - |
| Rat Genome U34 Set | L25331_at | - | - |
| Rat Genome U34 Set | rc_AA998747_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22150904 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358231 | - | - |
| RatRef-12 Expression BeadChip | ILMN_52418 | - | - |

Plod1
Genomic Map