Assay Details
| Gene Symbol: Kcnj9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053834.1 | NP_446286.1 | 2 - 3 | 223 | 123 | |
| GenBank mRNA | L77929.1 | - | 2 - 3 | 223 | 123 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the L77929.1, NM_053834.1 transcripts.
Set Membership: |
|

Kcnj9
Genomic Map