Assay Details
| Gene Symbol: Sdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053893.3 | NP_446345.3 | 4 - 5 | 1206 | 71 | |
| GenBank mRNA | U52825.1 | - | 4 - 5 | 1209 | 71 | |
| U73184.1 | - | 4 - 5 | 1274 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P109948 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P929767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311295 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662443 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716996 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28435 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P109948 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P929767 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P109948 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P929767 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P929767 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P109948 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12972 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 64135 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1109 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68118 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10880127 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880128 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880125 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880132 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880131 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880126 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880133 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880130 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880129 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880134 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10880124 | - | - |
| Rat Genome 230 2.0 Array | 1372180_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368580_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368581_at | - | - |
| Rat Genome 230 2.0 Array | 1368581_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368580_at | - | - |
| Rat Genome 230 2.0 Array | 1372180_at | - | - |
| Affymetrix HT Rat Focus Array | 1368581_at | - | - |
| Affymetrix HT Rat Focus Array | 1368581_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368580_at | - | - |
| Affymetrix HT Rat Focus Array | 1368580_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500011313.rn.1 | TC0500001567.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500011306.rn.1 | TC0500001567.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500011311.rn.1 | TC0500001567.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500011308.rn.1 | TC0500001567.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500011307.rn.1 | TC0500001567.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500011315.rn.1 | TC0500001567.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500011314.rn.1 | TC0500001567.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500011309.rn.1 | TC0500001567.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500011312.rn.1 | TC0500001567.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500011310.rn.1 | TC0500001567.rn.1 | RTA TC |
| Rat Expression Set 230 | 1372180_PM_at | - | - |
| Rat Expression Set 230 | 1372180_at | - | - |
| Rat Expression Set 230 | 1368580_PM_at | - | - |
| Rat Expression Set 230 | 1368581_PM_at | - | - |
| Rat Expression Set 230 | 1368580_at | - | - |
| Rat Expression Set 230 | 1368581_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880124 | - | - |
| Rat Genome U34 Set | X63143_s_at | - | - |
| Rat Genome U34 Set | rc_AI029642_at | - | - |
| Rat Genome U34 Set | rc_AA964067_at | - | - |
| Rat Genome U34 Set | X63143_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21353282 | - | - |
| ABI Rat Genome Survey Microarray | 21860762 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1354087 | - | - |
| RatRef-12 Expression BeadChip | ILMN_69620 | - | - |

Sdc3
Genomic Map