Assay Details
| Gene Symbol: Gp1bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB027146.1 | - | 1 | 753 | 55 | |
| Gene Symbol: Sept5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053931.3 | NP_446383.3 | 11 | 2759 | 55 | |
| GenBank mRNA | AB027143.1 | - | 11 | 2760 | 55 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P000539 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P124200 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P000534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33297 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P000539 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P124200 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P000534 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P000539 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P124200 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P000534 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P000539 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P124200 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P000534 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P151459 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P451244 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700886757 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 11350 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700886757 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10752288 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752276 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752279 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752285 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752288 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752294 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752286 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752287 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752287 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752291 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752284 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752275 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752294 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752282 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752280 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752278 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752277 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752274 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752290 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752272 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752273 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752290 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752291 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752281 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752293 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752293 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752283 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10752270 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10752289 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10752270 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10752289 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10752292 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10752292 | - | - |
| Rat Genome 230 2.0 Array | 1386955_at | - | - |
| Rat Genome 230 2.0 Array | 1367852_s_at | - | - |
| Rat Genome 230 2.0 Array | 1386955_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1386955_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1386955_at | - | - |
| Rat Genome 230 2.0 Array | 1367852_PM_s_at | - | - |
| Rat Genome 230 2.0 Array | 1367852_s_at | - | - |
| Rat Genome 230 2.0 Array | 1367852_PM_s_at | - | - |
| Affymetrix HT Rat Focus Array | 1367852_s_at | - | - |
| Affymetrix HT Rat Focus Array | 1367852_PM_s_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1100005600.rn.1 | TC1100000984.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100005601.rn.1 | TC1100000984.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100005603.rn.1 | TC1100000984.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100005604.rn.1 | TC1100000984.rn.1 | RTA TC |
| Rat Expression Set 230 | 1367852_s_at | - | - |
| Rat Expression Set 230 | 1367852_s_at | - | - |
| Rat Expression Set 230 | 1367852_PM_s_at | - | - |
| Rat Expression Set 230 | 1367852_PM_s_at | - | - |
| Rat Expression Set 230 | 1386955_at | - | - |
| Rat Expression Set 230 | 1386955_PM_at | - | - |
| Rat Expression Set 230 | 1386955_PM_at | - | - |
| Rat Expression Set 230 | 1386955_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752289 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752289 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752292 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752270 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752292 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752270 | - | - |
| Rat Genome U34 Set | rc_AA800851_s_at | - | - |
| Rat Genome U34 Set | rc_AA800851_s_at | - | - |
| Rat Genome U34 Set | rc_AA943092_at | - | - |
| Rat Genome U34 Set | rc_AA944408_g_at | - | - |
| Rat Genome U34 Set | rc_AA944408_at | - | - |
| Rat Genome U34 Set | rc_AA943092_at | - | - |
| Rat Genome U34 Set | rc_AA944408_g_at | - | - |
| Rat Genome U34 Set | rc_AA944408_at | - | - |
| Rat Genome U34 Set | rc_AI137922_at | - | - |
| Rat Genome U34 Set | rc_AI137922_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20944970 | - | - |
| ABI Rat Genome Survey Microarray | 21623054 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350620 | - | - |
| RatRef-12 Expression BeadChip | ILMN_59262 | - | - |

Gp1bb
Genomic Map