Assay Details
| Gene Symbol: Ccr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053958.1 | NP_446410.1 | 1 - 2 | 204 | 137 | |
| XM_006244178.2 | XP_006244240.1 | 1 - 2 | 982 | 137 | ||
| GenBank mRNA | Y13400.1 | - | 1 - 2 | 204 | 137 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_053958.1, XM_006244178.2, Y13400.1 transcripts.
Set Membership: |
|

Ccr3
Genomic Map