Assay Details
| Gene Symbol: Akap12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057103.2 | NP_476444.2 | 1 - 2 | 308 | 135 | |
| GenBank mRNA | AY695056.1 | - | 1 - 2 | 308 | 135 | |
| U41453.1 | - | 1 - 2 | 184 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Akap12
Genomic Map