Assay Details
| Gene Symbol: Nploc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080577.2 | NP_542144.1 | 4 - 5 | 541 | 86 | |
| GenBank mRNA | AF234600.1 | - | 4 - 5 | 469 | 86 | |
| BC101887.1 | - | 4 - 5 | 541 | 7930738 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P549675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386177 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267085 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130736 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P549675 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P549675 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P549675 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15352 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700330419 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92847 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68188 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 34696 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700330419 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10749633 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749646 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749643 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749635 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749640 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749642 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749632 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749644 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749638 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749647 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749631 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749639 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749636 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749634 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749641 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749645 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749637 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10749630 | - | - |
| Rat Genome 230 2.0 Array | 1374996_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374996_at | - | - |
| Rat Genome 230 2.0 Array | 1368929_at | - | - |
| Rat Genome 230 2.0 Array | 1393244_at | - | - |
| Rat Genome 230 2.0 Array | 1393244_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368929_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374996_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368929_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374996_at | - | - |
| Affymetrix HT Rat Focus Array | 1393244_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368929_at | - | - |
| Affymetrix HT Rat Focus Array | 1393244_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000038903.rn.1 | TC1000004265.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374996_PM_at | - | - |
| Rat Expression Set 230 | 1368929_PM_at | - | - |
| Rat Expression Set 230 | 1368929_at | - | - |
| Rat Expression Set 230 | 1374996_at | - | - |
| Rat Expression Set 230 | 1393244_PM_at | - | - |
| Rat Expression Set 230 | 1393244_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749630 | - | - |
| Rat Genome U34 Set | rc_AI045056_at | - | - |
| Rat Genome U34 Set | rc_AI172269_at | - | - |
| Rat Genome U34 Set | rc_AI138093_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21154733 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370237 | - | - |
| RatRef-12 Expression BeadChip | ILMN_67824 | - | - |

Nploc4
Genomic Map