Assay Details
| Gene Symbol: Dynll2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080697.2 | NP_542428.1 | 1 - 2 | 249 | 90 | |
| GenBank mRNA | AY034383.1 | - | 1 - 2 | 143 | 90 | |
| BC061874.1 | - | 1 - 2 | 249 | 5598327 | 90 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AY034383.1,BC061874.1,NM_080697.2 transcripts.
Set Membership: |
|

Dynll2
Genomic Map