Assay Details
| Gene Symbol: Psmb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080767.2 | NP_542945.2 | 1 - 2 | 198 | 98 | |
| GenBank mRNA | D10729.1 | - | 1 - 2 | 134 | 98 | |
| FQ232382.1 | - | 1 - 2 | 143 | 98 | ||
| FQ233197.1 | - | 1 - 2 | 176 | 98 | ||
| FQ234493.1 | - | 1 - 2 | 176 | 98 | ||
| FQ235267.1 | - | 1 - 2 | 187 | 98 | ||
| KC222892.1 | - | 1 - 2 | 148 | 98 | ||
| KC222893.1 | - | 1 - 2 | 148 | 98 | ||
| KC222894.1 | - | 1 - 2 | 148 | 98 | ||
| KC222895.1 | - | 1 - 2 | 148 | 98 | ||
| KC222896.1 | - | 1 - 2 | 148 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the D10729.1 transcripts.
Set Membership: |
|

Psmb8
Genomic Map