Assay Details
| Gene Symbol: Ifi27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203410.1 | NP_981955.1 | 3 - 4 | 327 | 81 | |
| GenBank mRNA | AF154572.1 | - | 2 - 3 | 251 | 81 | |
| BC060548.1 | - | 3 - 4 | 327 | 6919535 | 81 | |
| FQ219860.1 | - | 3 - 4 | 385 | 81 | ||
| FQ220098.1 | - | 3 - 4 | 396 | 81 | ||
| FQ226050.1 | - | 2 - 3 | 446 | 81 | ||
| FQ226998.1 | - | 3 - 4 | 360 | 81 | ||
| FQ230691.1 | - | 2 - 3 | 347 | 81 | ||
| FQ233730.1 | - | 3 - 4 | 361 | 81 | ||
| FQ233981.1 | - | 3 - 4 | 396 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ifi27
Genomic Map