Assay Details
| Gene Symbol: Tob1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133317.2 | NP_579851.2 | 1 | 342 | 63 | |
| GenBank mRNA | AF349723.1 | - | 1 | 337 | 63 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tob1
Genomic Map