Assay Details
| Gene Symbol: Cd79b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133533.2 | NP_598217.1 | 2 - 3 | 178 | 71 | |
| GenBank mRNA | BC088115.1 | - | 2 - 3 | 178 | 7325406 | 71 |
| FQ226266.1 | - | 2 - 3 | 175 | 71 | ||
| FQ227061.1 | - | 2 - 3 | 173 | 71 | ||
| FQ227788.1 | - | 2 - 3 | 167 | 71 | ||
| FQ230261.1 | - | 2 - 3 | 169 | 71 | ||
| FQ232054.1 | - | 2 - 3 | 169 | 71 | ||
| FQ232680.1 | - | 2 - 3 | 169 | 71 | ||
| FQ233507.1 | - | 2 - 3 | 167 | 71 | ||
| FQ234082.1 | - | 2 - 3 | 152 | 71 | ||
| FQ234487.1 | - | 2 - 3 | 167 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd79b
Genomic Map