Assay Details
| Gene Symbol: B3galt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133553.2 | NP_598237.2 | 1 | 384 | 125 | |
| GenBank mRNA | AB003478.1 | - | 1 | 355 | 125 | |
| BC126083.1 | - | 1 | 384 | 8375818 | 125 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

B3galt4
Genomic Map