Assay Details
| Gene Symbol: Cdc25b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133572.1 | NP_598256.1 | 3 - 4 | 612 | 65 | |
| XM_006235003.1 | XP_006235065.1 | 3 - 4 | 585 | 65 | ||
| GenBank mRNA | D16237.1 | - | 3 - 4 | 612 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdc25b
Genomic Map